Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Phenylcyclohexanone, 98%

Catalog No. AAA1734006
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1734006 5 g
AAA1734003 1 g
AAA1734014 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1734006 Supplier Thermo Scientific Chemicals Supplier No. A1734006
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

CAS: 1444-65-1 | C12H14O | 174.243 g/mol

It can be used to produce 6-oxo-6-phenyl-hexanoic acid.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It can be used to produce 6-oxo-6-phenyl-hexanoic acid.

Solubility
Slightly soluble in water.

Notes
Storage temperature: 2 - 4°Cand Keep container tightly closed in a dryand well-ventilated place. Keep away from strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1444-65-1
Molecular Formula C12H14O
Molecular Weight (g/mol) 174.243
MDL Number MFCD00001629
InChI Key DRLVMOAWNVOSPE-UHFFFAOYSA-N
Synonym 2-phenylcyclohexanone, cyclohexanone, 2-phenyl, 2-phenyl-cyclohexanone, 2-phenyl cyclohexanone, cyclohexanone, phenyl, 2-phenylcyclohexanon, phenylcyclohexanone, phenyl cyclohexanone, 6-phenylcyclohexanone
PubChem CID 95592
IUPAC Name 2-phenylcyclohexan-1-one
SMILES C1CCC(=O)C(C1)C2=CC=CC=C2

Specifications

Melting Point 56°C to 59°C
Quantity 5 g
Solubility Information Slightly soluble in water.
Formula Weight 174.24
Percent Purity 98%
Chemical Name or Material 2-Phenylcyclohexanone
EINECSNumber 215-888-7
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.