Learn More
Medchemexpress LLC 2-Phenylethanol | 60-12-8 | 100.0% | 250 G
SDP
Catalog No. 5000413946
Encompass_Preferred
2-Phenylethanol is an aromatic alcohol with a rose-like odor, commonly used as a flavor and fragrance compound, preservative, and anti-microbial agent. It exhibits antityrosinase and antimicrobial activities, inhibiting the growth of Gram-negative microorganisms. In vitro, it is used as an additive in cigarettes and a preservative in soaps. It appears as a colorless to light yellow liquid with a density of 1.020 g/cm3, has a molecular weight of 122.16 and its formula is C8H10O.
  • Aromatic alcohol with a rose-like odor
  • Used as a flavor and fragrance compound
  • Acts as a preservative and anti-microbial agent
  • Exhibits antityrosinase and antimicrobial activities
  • Inhibits growth of gram-negative microorganisms
  • Colorless to light yellow liquid
Catalog No. 50-004-13946 Supplier Medchemexpress LLC Supplier No. 250GHYB1290
2-Phenylethanol is an aromatic alcohol with a rose-like odor, commonly used as a flavor and fragrance compound, preservative, and anti-microbial agent. It exhibits antityrosinase and antimicrobial activities, inhibiting the growth of Gram-negative microorganisms. In vitro, it is used as an additive in cigarettes and a preservative in soaps. It appears as a colorless to light yellow liquid with a density of 1.020 g/cm3, has a molecular weight of 122.16 and its formula is C8H10O.
  • Aromatic alcohol with a rose-like odor
  • Used as a flavor and fragrance compound
  • Acts as a preservative and anti-microbial agent
  • Exhibits antityrosinase and antimicrobial activities
  • Inhibits growth of gram-negative microorganisms
  • Colorless to light yellow liquid