Learn More
Medchemexpress LLC 2-Phenylethanol | 60-12-8 | 99.9% | 100 G
SDP
Catalog No. 5000412503
Encompass_Preferred
2-Phenylethanol is an aromatic alcohol characterized by a rose-like odor. It functions as a flavor and fragrance compound and can be utilized as a preservative and antimicrobial agent. This compound also demonstrates antityrosinase activity.
  • An aromatic alcohol with a rose-like odor
  • Functions as a flavor and fragrance compound
  • Can be used as a preservative
  • Acts as an antimicrobial agent
  • Exhibits antityrosinase activity
  • Inhibits the growth of Gram-negative microorganisms
Catalog No. 50-004-12503 Supplier Medchemexpress LLC Supplier No. HYB1290100G
2-Phenylethanol is an aromatic alcohol characterized by a rose-like odor. It functions as a flavor and fragrance compound and can be utilized as a preservative and antimicrobial agent. This compound also demonstrates antityrosinase activity.
  • An aromatic alcohol with a rose-like odor
  • Functions as a flavor and fragrance compound
  • Can be used as a preservative
  • Acts as an antimicrobial agent
  • Exhibits antityrosinase activity
  • Inhibits the growth of Gram-negative microorganisms