Learn More
Medchemexpress LLC 2-Phenylethanol | 60-12-8 | 99.99% | 1000 G
SDP
Catalog No. 5000412070
Encompass_Preferred
2-Phenylethanol, also known as phenethyl alcohol, is an aromatic alcohol recognized for its rose-like odor. It serves as a flavor and fragrance compound and is effective as a preservative and anti-microbial agent. This compound demonstrates both antityrosinase and antimicrobial activities, making it suitable for various applications.
  • Functions as a flavor and fragrance compound.
  • Acts as an effective preservative.
  • Demonstrates anti-microbial properties.
  • Exhibits antityrosinase activity.
  • Used as an additive in cigarettes and a preservative in soaps.
  • Inhibits the growth of Gram-negative microorganisms.
Catalog No. 50-004-12070 Supplier Medchemexpress LLC Supplier No. HYB12901000G
2-Phenylethanol, also known as phenethyl alcohol, is an aromatic alcohol recognized for its rose-like odor. It serves as a flavor and fragrance compound and is effective as a preservative and anti-microbial agent. This compound demonstrates both antityrosinase and antimicrobial activities, making it suitable for various applications.
  • Functions as a flavor and fragrance compound.
  • Acts as an effective preservative.
  • Demonstrates anti-microbial properties.
  • Exhibits antityrosinase activity.
  • Used as an additive in cigarettes and a preservative in soaps.
  • Inhibits the growth of Gram-negative microorganisms.