Learn More
Medchemexpress LLC 2-Phenylethanol | 60-12-8 | MFCD00002886 | 99.9% | 500 G
SDP
Catalog No. 5000413257
Encompass_Preferred
2-Phenylethanol (Phenethyl alcohol) is an aromatic alcohol with a rose-like odor. It is a flavor and fragrance compound and can be used as a preservative and anti-microbial agent. It also exhibits antityrosinase and antimicrobial activities. In vitro, it is used as an additive in cigarettes and a preservative in soaps, and it inhibits the growth of Gram-negative microorganisms.
  • Aromatic alcohol with a rose-like odor
  • Flavor and fragrance compound
  • Can be used as a preservative and anti-microbial agent
  • Exhibits antityrosinase and antimicrobial activities
  • Inhibitory effect on the growth of Gram-negative microorganisms
Catalog No. 50-004-13257 Supplier Medchemexpress LLC Supplier No. HYB1290500G
2-Phenylethanol (Phenethyl alcohol) is an aromatic alcohol with a rose-like odor. It is a flavor and fragrance compound and can be used as a preservative and anti-microbial agent. It also exhibits antityrosinase and antimicrobial activities. In vitro, it is used as an additive in cigarettes and a preservative in soaps, and it inhibits the growth of Gram-negative microorganisms.
  • Aromatic alcohol with a rose-like odor
  • Flavor and fragrance compound
  • Can be used as a preservative and anti-microbial agent
  • Exhibits antityrosinase and antimicrobial activities
  • Inhibitory effect on the growth of Gram-negative microorganisms