Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 2-Phenylethanol-d5 | 35845-63-7 | 10 MG
SDP

Catalog No. 5000297400
Encompass_Preferred

2-Phenylethanol-d5 is the deuterium-labeled analogue of 2-phenylethanol supplied for analytical and research use. It preserves the parent compound's chemical properties while producing a mass/spectral shift useful for tracing and quantitative assays.

  • Deuterium-labeled internal standard for quantitative analysis.
  • Suitable for NMR, GC-MS, and LC-MS applications.
  • Available in small research-scale pack sizes such as 10 mg.
  • Molecular formula C8H5D5O; molecular weight 127.20 g·mol⁻¹.
  • CAS number 35845-63-7 for unambiguous identification.

Catalog No. 50-002-97400 Supplier Medchemexpress LLC Supplier No. HYB1290S210MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

2-Phenylethanol-d5 is the deuterium-labeled analogue of 2-phenylethanol supplied for analytical and research use. It preserves the parent compound's chemical properties while producing a mass/spectral shift useful for tracing and quantitative assays.

  • Deuterium-labeled internal standard for quantitative analysis.
  • Suitable for NMR, GC-MS, and LC-MS applications.
  • Available in small research-scale pack sizes such as 10 mg.
  • Molecular formula C8H5D5O; molecular weight 127.20 g·mol⁻¹.
  • CAS number 35845-63-7 for unambiguous identification.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.