Learn More
Medchemexpress LLC 2-Phenylethanol-d5 | 35845-63-7 | 98.3% | 25 MG
SDP
Catalog No. 5000414417
Encompass_Preferred
2-Phenylethanol-d5 is a deuterium-labeled form of 2-Phenylethanol, a colorless liquid with a pleasant floral odor. This compound is naturally extracted from various plants like rose, carnation, hyacinth, Aleppo pine, and orange blossom. It also acts as an autoantibiotic produced by the fungus Candida albicans. Its applications include being an additive in cigarettes and a preservative in soaps due to its stability in basic conditions.
  • Deuterium-labeled compound.
  • Colorless liquid with a pleasant floral odor.
  • Naturally occurring in several plant species.
  • Exhibits autoantibiotic properties from Candida albicans.
  • Used as an additive in cigarettes.
  • Functions as a preservative in soaps due to high stability.
Catalog No. 50-004-14417 Supplier Medchemexpress LLC Supplier No. 25MGHYB1290S2
2-Phenylethanol-d5 is a deuterium-labeled form of 2-Phenylethanol, a colorless liquid with a pleasant floral odor. This compound is naturally extracted from various plants like rose, carnation, hyacinth, Aleppo pine, and orange blossom. It also acts as an autoantibiotic produced by the fungus Candida albicans. Its applications include being an additive in cigarettes and a preservative in soaps due to its stability in basic conditions.
  • Deuterium-labeled compound.
  • Colorless liquid with a pleasant floral odor.
  • Naturally occurring in several plant species.
  • Exhibits autoantibiotic properties from Candida albicans.
  • Used as an additive in cigarettes.
  • Functions as a preservative in soaps due to high stability.