Learn More
Medchemexpress LLC 2-Phenylethanol-d5 | 35845-63-7 | 98.3% | 5 MG
SDP
Catalog No. 5000413613
Encompass_Preferred
2-Phenylethanol-d5 is a deuterium-labeled variant of 2-Phenylethanol. This colorless liquid is naturally extracted from various botanicals such as rose, carnation, hyacinth, Aleppo pine, and orange blossom. It is recognized for its pleasant floral odor and also functions as an autoantibiotic produced by the fungus Candida albicans. Its deuterated form is used as a tracer and internal standard in drug development and analysis.
  • Used as an additive in cigarettes.
  • Functions as a preservative in soaps due to its stability in basic conditions.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
  • Deuteration has potential to influence the pharmacokinetic and metabolic profiles of drugs.
Catalog No. 50-004-13613 Supplier Medchemexpress LLC Supplier No. HYB1290S25MG
2-Phenylethanol-d5 is a deuterium-labeled variant of 2-Phenylethanol. This colorless liquid is naturally extracted from various botanicals such as rose, carnation, hyacinth, Aleppo pine, and orange blossom. It is recognized for its pleasant floral odor and also functions as an autoantibiotic produced by the fungus Candida albicans. Its deuterated form is used as a tracer and internal standard in drug development and analysis.
  • Used as an additive in cigarettes.
  • Functions as a preservative in soaps due to its stability in basic conditions.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
  • Deuteration has potential to influence the pharmacokinetic and metabolic profiles of drugs.