Learn More
Medchemexpress LLC 2-Phenylethanol (Standard) | 60-12-8 | 100.0% | 100 MG
SDP
Catalog No. 5000412071
Encompass_Preferred
2-Phenylethanol is the analytical standard for 2-Phenylethanol, intended for research and analytical applications. Also known as Phenethyl alcohol, it is an aromatic alcohol with a rose-like odor. It functions as a flavor and fragrance compound, preservative, and anti-microbial agent, exhibiting antityrosinase and antimicrobial activities.
  • Analytical standard for 2-phenylethanol
  • Intended for research and analytical applications
  • Aromatic alcohol with a rose-like odor
  • Flavor and fragrance compound
  • Preservative and anti-microbial agent
  • Exhibits antityrosinase and antimicrobial activities
  • Used in qualitative, quantitative, and methodological research experiments in HPLC, GC, and MS
Catalog No. 50-004-12071 Supplier Medchemexpress LLC Supplier No. HYB1290R100MG
2-Phenylethanol is the analytical standard for 2-Phenylethanol, intended for research and analytical applications. Also known as Phenethyl alcohol, it is an aromatic alcohol with a rose-like odor. It functions as a flavor and fragrance compound, preservative, and anti-microbial agent, exhibiting antityrosinase and antimicrobial activities.
  • Analytical standard for 2-phenylethanol
  • Intended for research and analytical applications
  • Aromatic alcohol with a rose-like odor
  • Flavor and fragrance compound
  • Preservative and anti-microbial agent
  • Exhibits antityrosinase and antimicrobial activities
  • Used in qualitative, quantitative, and methodological research experiments in HPLC, GC, and MS