Learn More
Medchemexpress LLC 2-Phenylethanol (Standard) | 60-12-8 | 99.99% | 25 MG
SDP
Catalog No. 5000414276
Encompass_Preferred
The 2-Phenylethanol (Standard) is an analytical standard of 2-Phenylethanol, an aromatic alcohol that has a rose-like odor and is used as a flavor and fragrance compound. It also acts as a preservative and anti-microbial agent, exhibiting antityrosinase and antimicrobial activities. This product is intended for research and analytical applications, commonly utilized in qualitative, quantitative, and methodological research experiments such as HPLC, GC, and MS.
  • Analytical standard for research and analytical applications.
  • Used in qualitative, quantitative, and methodological research experiments.
  • Acts as a flavor and fragrance compound.
  • Functions as a preservative.
  • Serves as an anti-microbial agent.
  • Exhibits antityrosinase activities.
  • Demonstrates antimicrobial activities.
Catalog No. 50-004-14276 Supplier Medchemexpress LLC Supplier No. 25MGHYB1290R
The 2-Phenylethanol (Standard) is an analytical standard of 2-Phenylethanol, an aromatic alcohol that has a rose-like odor and is used as a flavor and fragrance compound. It also acts as a preservative and anti-microbial agent, exhibiting antityrosinase and antimicrobial activities. This product is intended for research and analytical applications, commonly utilized in qualitative, quantitative, and methodological research experiments such as HPLC, GC, and MS.
  • Analytical standard for research and analytical applications.
  • Used in qualitative, quantitative, and methodological research experiments.
  • Acts as a flavor and fragrance compound.
  • Functions as a preservative.
  • Serves as an anti-microbial agent.
  • Exhibits antityrosinase activities.
  • Demonstrates antimicrobial activities.