Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Phenylethyl-1-boronic acid pinacol ester, 99%

Catalog No. AAL1953003
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1953003 1 g
AAL19530MD 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL1953003 Supplier Thermo Scientific Chemicals Supplier No. L1953003
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

CAS: 165904-22-3 | C14H21BO2 | 232.13 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 165904-22-3
Molecular Formula C14H21BO2
Molecular Weight (g/mol) 232.13
MDL Number MFCD03788721
InChI Key LVLQNRWCBBIVHR-UHFFFAOYSA-N
Synonym 4,4,5,5-tetramethyl-2-phenethyl-1,3,2-dioxaborolane, 2-phenylethyl-1-boronic acid pinacol ester, 4,4,5,5-tetramethyl-2-2-phenylethyl-1,3,2-dioxaborolane, 2-phenylethylboronic acid, pinacol ester, phenethylboronic acid pinacol ester, 2-phenylethylboronic acid pinacol ester, 2-phenylethylboronic acid,pinacol ester, 2-phenylethyl boronic acid pinacol ester, 2-phenyl ethyl-1-boronic acid pinacol ester, 2-phenethyl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
PubChem CID 11333720
IUPAC Name 4,4,5,5-tetramethyl-2-(2-phenylethyl)-1,3,2-dioxaborolane
SMILES B1(OC(C(O1)(C)C)(C)C)CCC2=CC=CC=C2

Specifications

Melting Point 31°C to 33°C
Boiling Point 70°C to 80°C
Refractive Index 1.486
Quantity 1 g
Beilstein 8258927
Formula Weight 232.13
Percent Purity 99%
Chemical Name or Material 2-Phenylethyl-1-boronic acid pinacol ester
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.