Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 2-Phenylethyl β-D-thiogalactoside | 63407-54-5 | 99.9% | 300.37 | 250 MG
SDP

Catalog No. 5000350157
Encompass_Preferred

2-Phenylethyl β-D-thiogalactoside is a biochemical reagent designed for life science research. This compound appears as a white to off-white solid, possesses a molecular weight of 300.37, and exhibits high purity. It is stable under appropriate storage conditions, making it suitable for various laboratory applications.

  • Suitable for life science-related research
  • Can be utilized as a biological material or organic compound
  • Purity of 99.9%
  • Solid appearance, white to off-white
  • Store solid at 4°C under nitrogen
  • Store in solvent at -80°C for 6 months, or -20°C for 1 month under nitrogen

Catalog No. 50-003-50157 Supplier Medchemexpress LLC Supplier No. 250MGHY137201
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

2-Phenylethyl β-D-thiogalactoside is a biochemical reagent designed for life science research. This compound appears as a white to off-white solid, possesses a molecular weight of 300.37, and exhibits high purity. It is stable under appropriate storage conditions, making it suitable for various laboratory applications.

  • Suitable for life science-related research
  • Can be utilized as a biological material or organic compound
  • Purity of 99.9%
  • Solid appearance, white to off-white
  • Store solid at 4°C under nitrogen
  • Store in solvent at -80°C for 6 months, or -20°C for 1 month under nitrogen

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.