Learn More
Sigma Aldrich Fine Chemicals Biosciences 2 Phenylethyl isothiocyana1KG
Catalog No. 502907965
Encompass
2-Phenylethyl isothiocyanate (2-PE ITC) is a bioactive isothiocyanate mainly found in Brassica vegetables. It is produced by the enzymatic hydrolysis of 2-phenylethyl glucosinolate. 2-PE ITC is the main compound responsible for the nematicidal effect of Brassica root tissues against (Pratylenchus neglectus Filipjev) a root-lesion nematode. This ability makes it a promising candidate for the development of soil fumigants.
Catalog No. 50-290-7965 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. W4014041KGK
2-Phenylethyl isothiocyanate (2-PE ITC) is a bioactive isothiocyanate mainly found in Brassica vegetables. It is produced by the enzymatic hydrolysis of 2-phenylethyl glucosinolate. 2-PE ITC is the main compound responsible for the nematicidal effect of Brassica root tissues against (Pratylenchus neglectus Filipjev) a root-lesion nematode. This ability makes it a promising candidate for the development of soil fumigants.