Learn More
Medchemexpress LLC 2-Phenylethyl isothiocyanate (2-PE ITC) | 2257-09-2 | 98.00% | 163.24 | 100 MG
SDP
Catalog No. 5000401946
Encompass_Preferred
2-Phenylethyl isothiocyanate is a potent antifungal agent that significantly inhibits spore germination and mycelial growth of *Alternaria alternata* with a minimum inhibitory concentration (MIC) of 1.22 mM. Its antifungal effect against *Alternaria alternata* may involve reducing toxin content and breaking down cell membrane integrity.
  • Significantly inhibited spore germination.
  • Significantly inhibited mycelial growth.
  • Reduces toxin content.
  • Breaks down cell membrane integrity.
Catalog No. 50-004-01946 Supplier Medchemexpress LLC Supplier No. HY23155100MG
2-Phenylethyl isothiocyanate is a potent antifungal agent that significantly inhibits spore germination and mycelial growth of *Alternaria alternata* with a minimum inhibitory concentration (MIC) of 1.22 mM. Its antifungal effect against *Alternaria alternata* may involve reducing toxin content and breaking down cell membrane integrity.
  • Significantly inhibited spore germination.
  • Significantly inhibited mycelial growth.
  • Reduces toxin content.
  • Breaks down cell membrane integrity.