Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 2-Phenylethyl isothiocyanate | 2257-09-2 | 98.0% | 163.24 | 1 ML
SDP

Catalog No. 5000400838
Encompass_Preferred

2-Phenylethyl isothiocyanate | 2257-09-2 | 98.0% | 163.24 | 1 ML

Catalog No. 50-004-00838 Supplier Medchemexpress LLC Supplier No. HY2315510MM/1ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

2-Phenylethyl isothiocyanate | 2257-09-2 | 98.0% | 163.24 | 1 ML

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.