Learn More
Medchemexpress LLC 2-Phenylethyl isothiocyanate | 2257-09-2 | 98.0% | 163.24 | 5 MG
SDP
Catalog No. 5000400855
Encompass_Preferred
2-Phenylethyl isothiocyanate (2-PE ITC) is described as a potent antifungal agent. It has been shown to significantly inhibit spore germination and mycelial growth of Alternaria alternata, with a minimum inhibitory concentration (MIC) of 1.22 mM. The antifungal effects against Alternaria alternata may be due to a reduction in toxin content and the breakdown of cell membrane integrity. For research use only, this product is not sold to patients. It significantly controls black spot caused by A. alternata and damages membrane integrity of A. alternata in pear fruit.
  • Potent antifungal agent
  • Significantly inhibits spore germination and mycelial growth of Alternaria alternata
  • Reduces toxin content in Alternaria alternata
  • Breaks down cell membrane integrity in Alternaria alternata
  • Controls black spot in pear fruit
Catalog No. 50-004-00855 Supplier Medchemexpress LLC Supplier No. HY23155S5MG
2-Phenylethyl isothiocyanate (2-PE ITC) is described as a potent antifungal agent. It has been shown to significantly inhibit spore germination and mycelial growth of Alternaria alternata, with a minimum inhibitory concentration (MIC) of 1.22 mM. The antifungal effects against Alternaria alternata may be due to a reduction in toxin content and the breakdown of cell membrane integrity. For research use only, this product is not sold to patients. It significantly controls black spot caused by A. alternata and damages membrane integrity of A. alternata in pear fruit.
  • Potent antifungal agent
  • Significantly inhibits spore germination and mycelial growth of Alternaria alternata
  • Reduces toxin content in Alternaria alternata
  • Breaks down cell membrane integrity in Alternaria alternata
  • Controls black spot in pear fruit