Learn More
Selleck Chemical LLC 2-Phenylethylamine S5347-25ul
SDP
Catalog No. 503111519
Encompass_Preferred
2-Phenylethylamine ( -Phenylethylamine benzeneethanamine -aminoethylbenzene) is a natural monoamine alkaloid that functions as a monoaminergic neuromodulator or a neurotransmitter in the human central nervous system
Catalog No. 50-311-1519 Supplier Selleck Chemical LLC Supplier No. S534725UL
2-Phenylethylamine ( -Phenylethylamine benzeneethanamine -aminoethylbenzene) is a natural monoamine alkaloid that functions as a monoaminergic neuromodulator or a neurotransmitter in the human central nervous system