Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich 2-Phenylethylamine hydrochloride

Catalog No. P6513100G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P6513100G 100 g
P651325G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P6513100G Supplier Sigma Aldrich Supplier No. P6513100G
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

2-Phenylethylamine hydrochloride

Specifications

CAS 156-28-5
Melting Point 220°C to 222°C (lit.)
Molecular Formula C8H11N -+ HCl
Linear Formula C6H5CH2CH2NH2 -+ HCl
MDL Number MFCD00012893
Quantity 100 g
Synonym beta-Phenylethylamine hydrochloride; Phenethylamine hydrochloride
Molecular Weight (g/mol) 157.64
Percent Purity ≥98%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.