Learn More
Medchemexpress LLC 2-Phenylglycine-d5 | 358731-96-1 | 99.0% | 156.19 | 10 MG
SDP
Catalog No. 5000452531
Encompass_Preferred
2-Phenylglycine-d5 is a deuterium-labeled version of 2-Phenylglycine. 2-Phenylglycine (DL-α-Phenylglycine) is a metabolite found in breast milk during the W2 to W4 lactation period.
  • Can be utilized as a tracer in research.
  • Can serve as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
  • Stable heavy isotopes like deuterium are incorporated into drug molecules, primarily as tracers for quantitation during the drug development process. Deuteration can impact the pharmacokinetic and metabolic profiles of drugs.
Catalog No. 50-004-52531 Supplier Medchemexpress LLC Supplier No. HYW010248S10MG
2-Phenylglycine-d5 is a deuterium-labeled version of 2-Phenylglycine. 2-Phenylglycine (DL-α-Phenylglycine) is a metabolite found in breast milk during the W2 to W4 lactation period.
  • Can be utilized as a tracer in research.
  • Can serve as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
  • Stable heavy isotopes like deuterium are incorporated into drug molecules, primarily as tracers for quantitation during the drug development process. Deuteration can impact the pharmacokinetic and metabolic profiles of drugs.