Learn More
Medchemexpress LLC 2-Phenylglycine-d5 (DL-α-Phenylglycine-d5) | 358731-96-1 | 99.0% | 156.19 | 5 MG
SDP
Catalog No. 5000453983
Encompass_Preferred
2-Phenylglycine-d5 is the deuterium labeled 2-Phenylglycine. It is intended for research use only. This compound is a metabolite found in breast milk during the W2 to W4 lactation period. Deuteration has gained attention for its potential to influence the pharmacokinetic and metabolic profiles of drugs.
  • Deuterium labeled 2-Phenylglycine
  • Metabolite found in breast milk during the W2 to W4 lactation period
  • Can be used as a tracer
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
Catalog No. 50-004-53983 Supplier Medchemexpress LLC Supplier No. HYW010248S5MG
2-Phenylglycine-d5 is the deuterium labeled 2-Phenylglycine. It is intended for research use only. This compound is a metabolite found in breast milk during the W2 to W4 lactation period. Deuteration has gained attention for its potential to influence the pharmacokinetic and metabolic profiles of drugs.
  • Deuterium labeled 2-Phenylglycine
  • Metabolite found in breast milk during the W2 to W4 lactation period
  • Can be used as a tracer
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS