Learn More
Medchemexpress LLC 2-phenylhydroquinone | 1079-21-6 | MFCD00002342 | 99.2% | 186.21 | C12H10O2 | 100 G
SDP
Catalog No. 5000302135
Encompass_Preferred
Phenylhydroquinone is a small-molecule research reagent reported to induce thymic atrophy and to up-regulate apoptosis through activation of p53. It is used in biochemical and cell-based studies to probe MDM2/p53, ERK, and p38 MAPK signaling pathways.
  • Induces thymic atrophy and modulates apoptosis pathways.
  • Reported activity against MDM2/p53, ERK, and p38 MAPK.
  • High reported purity suitable for research use.
  • Available in multiple package sizes, including 100 g.
  • Molecular formula C12H10O2 and molecular weight 186.21.
Catalog No. 50-003-02135 Supplier Medchemexpress LLC Supplier No. HYW016181100G
Phenylhydroquinone is a small-molecule research reagent reported to induce thymic atrophy and to up-regulate apoptosis through activation of p53. It is used in biochemical and cell-based studies to probe MDM2/p53, ERK, and p38 MAPK signaling pathways.
  • Induces thymic atrophy and modulates apoptosis pathways.
  • Reported activity against MDM2/p53, ERK, and p38 MAPK.
  • High reported purity suitable for research use.
  • Available in multiple package sizes, including 100 g.
  • Molecular formula C12H10O2 and molecular weight 186.21.