Learn More
Medchemexpress LLC 2-phenylimidazole | 670-96-2 | 99.2% | 144.18 | C9H8N2 | 500g
SDP
Catalog No. 5000267192
Encompass_Preferred
2-Phenylimidazole (PhI) is a corrosion inhibitor for brass and can be used to prepare complexes with ruthenium (III)[1]
Catalog No. 50-002-67192 Supplier Medchemexpress LLC Supplier No. HYY1312500G
2-Phenylimidazole (PhI) is a corrosion inhibitor for brass and can be used to prepare complexes with ruthenium (III)[1]