Learn More
Medchemexpress LLC 2-phenylimidazole | 670-96-2 | MFCD00005186 | 99.2% | 100g
SDP
Supplier:  Medchemexpress LLC HYY1312100G
Encompass_Preferred
2-Phenylimidazole (PhI) is a corrosion inhibitor for brass and can be used to prepare complexes with ruthenium (III)[1]
Catalog No. 50-002-67894