Learn More
Selleck Chemical LLC 2-Phenylimidazole
SDP
Catalog No. 5000003486
Encompass_Preferred
2-phenylimidazole (PhI 2-Phenyl-1H-imidazole) is a corrosion inhibitor for brass and can be used to prepare complex compounds with ruthenium(III)
Catalog No. 50-000-03486 Supplier Selleck Chemical LLC Supplier No. S587725MG
2-phenylimidazole (PhI 2-Phenyl-1H-imidazole) is a corrosion inhibitor for brass and can be used to prepare complex compounds with ruthenium(III)