Learn More
Medchemexpress LLC 2-phenylmelatonin | 151889-03-1 | 99.2% | 308.37 g/mol | C19H20N2O2 | 25 MG
SDP
Catalog No. 5000213482
Encompass_Preferred
UCM 608 (2-Phenylmelatonin) is a potent, high-affinity melatonin (MT) membrane receptor agonist provided as a purified small-molecule standard for in vitro pharmacology and assay development. It is intended for research use only and has documented binding data for MT1 and MT2.
  • High affinity for MT1 and MT2 (pKi 10.7 and 10.4).
  • High purity (99.18%).
  • Well characterized small molecule with formula C19H20N2O2 and molecular weight 308.37 g/mol.
  • Supplied in small quantities and solution formulations suitable for assays.
  • Intended for research use only; not for human or veterinary use.
Catalog No. 50-002-13482 Supplier Medchemexpress LLC Supplier No. HY10107425MG
UCM 608 (2-Phenylmelatonin) is a potent, high-affinity melatonin (MT) membrane receptor agonist provided as a purified small-molecule standard for in vitro pharmacology and assay development. It is intended for research use only and has documented binding data for MT1 and MT2.
  • High affinity for MT1 and MT2 (pKi 10.7 and 10.4).
  • High purity (99.18%).
  • Well characterized small molecule with formula C19H20N2O2 and molecular weight 308.37 g/mol.
  • Supplied in small quantities and solution formulations suitable for assays.
  • Intended for research use only; not for human or veterinary use.