Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 2-phenylmelatonin | 151889-03-1 | 99.2% | 308.37 g/mol | C19H20N2O2 | 5 MG
SDP

Catalog No. 5000213298
Encompass_Preferred

2-Phenylmelatonin is a high-affinity melatonin membrane receptor agonist used for research applications. It demonstrates potent binding to MT1 and MT2 receptors and is supplied as a high-purity powder suitable for in vitro and in vivo studies.

  • High affinity for MT1 and MT2 receptors (pKi 10.7, 10.4).
  • High purity (~99.2%).
  • Molecular weight 308.37 g/mol.
  • Available in small research quantities (for example, 5 mg).
  • Soluble in DMSO (50 mg/mL) and compatible with common in vivo formulations.
  • Stable as powder when stored at -20°C for long-term storage.
  • Intended for research use only.

Catalog No. 50-002-13298 Supplier Medchemexpress LLC Supplier No. HY1010745MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

2-Phenylmelatonin is a high-affinity melatonin membrane receptor agonist used for research applications. It demonstrates potent binding to MT1 and MT2 receptors and is supplied as a high-purity powder suitable for in vitro and in vivo studies.

  • High affinity for MT1 and MT2 receptors (pKi 10.7, 10.4).
  • High purity (~99.2%).
  • Molecular weight 308.37 g/mol.
  • Available in small research quantities (for example, 5 mg).
  • Soluble in DMSO (50 mg/mL) and compatible with common in vivo formulations.
  • Stable as powder when stored at -20°C for long-term storage.
  • Intended for research use only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.