Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 2-phenylmelatonin | 151889-03-1 | MFCD00673896 | 99.2% | 308.37 g/mol | C19H20N2O2 | 10 MG
SDP

Supplier:  Medchemexpress LLC HY10107410MG

Encompass_Preferred

UCM 608 (2-Phenylmelatonin) is a high-affinity melatonin (MT) receptor agonist supplied as a purified solid for laboratory research. It binds MT1 and MT2 receptors with reported pKi values of 10.7 and 10.4, respectively, and is used in pharmacology and receptor-binding studies. Handle and store according to safety data and laboratory protocols.

  • High affinity for MT1 and MT2 receptors.
  • Reported pKi values: MT1 10.7, MT2 10.4.
  • Molecular formula C19H20N2O2.
  • Molecular weight 308.37 g/mol.
  • Purity approximately 99.18% (reported).
  • Solid, off-white to yellow appearance.
  • Store powder at -20°C or 4°C per stability guidance.
  • Intended for research use only.

Catalog No. 50-002-13251


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.