Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 2-phenylmelatonin | 151889-03-1 | MFCD00673896 | 99.2% | 308.37 g/mol | C19H20N2O2 | 50 MG
SDP

Catalog No. 5000213395
Encompass_Preferred

2-phenylmelatonin is a high-affinity melatonin membrane receptor agonist used in receptor pharmacology studies; it displays pKi values of 10.7 (MT1) and 10.4 (MT2) and is supplied as a high-purity solid for research applications.

  • High-affinity melatonin receptor agonist with pKi MT1 10.7 and MT2 10.4.
  • Molecular formula C19H20N2O2 and molecular weight 308.37 g/mol.
  • CAS number 151889-03-1 for unambiguous identification.
  • High purity (99.18%) solid suitable for in vitro and in vivo studies.
  • Soluble in DMSO at 50 mg/mL; in vivo formulations reported at ≥1 mg/mL with common vehicles.
  • Store powder at -20°C for long-term stability; store solutions at -80°C when indicated.

Catalog No. 50-002-13395 Supplier Medchemexpress LLC Supplier No. HY10107450MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

2-phenylmelatonin is a high-affinity melatonin membrane receptor agonist used in receptor pharmacology studies; it displays pKi values of 10.7 (MT1) and 10.4 (MT2) and is supplied as a high-purity solid for research applications.

  • High-affinity melatonin receptor agonist with pKi MT1 10.7 and MT2 10.4.
  • Molecular formula C19H20N2O2 and molecular weight 308.37 g/mol.
  • CAS number 151889-03-1 for unambiguous identification.
  • High purity (99.18%) solid suitable for in vitro and in vivo studies.
  • Soluble in DMSO at 50 mg/mL; in vivo formulations reported at ≥1 mg/mL with common vehicles.
  • Store powder at -20°C for long-term stability; store solutions at -80°C when indicated.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.