Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Phenylnaphthalene 95.0+%, TCI America™
SDP

Catalog No. P06791G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P06791G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. P06791G Supplier TCI America Supplier No. P06791G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 612-94-2
Molecular Formula C16H12
Molecular Weight (g/mol) 204.272
MDL Number MFCD00039601
InChI Key TURIHPLQSRVWHU-UHFFFAOYSA-N
Synonym naphthalene, 2-phenyl, .beta.-phenylnaphthalene, unii-949vn5dh7a, beta-phenylnaphthalene, 6-phenylnaphthalene, 2-phenyl-naphthalene, acmc-1bas3, naphthalene, 2-phenyl-8ci 9ci, 10 microg/ml in cyclohexane
PubChem CID 11934
IUPAC Name 2-phenylnaphthalene
SMILES C1=CC=C(C=C1)C2=CC3=CC=CC=C3C=C2

Specifications

Melting Point 105°C
Color White
Boiling Point 358°C
Quantity 1 g
Formula Weight 204.27
Percent Purity ≥95.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2-Phenylnaphthalene
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.