Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Phenylphenol, 99+%

Catalog No. p-183398
Change view
Click to view available options
Quantity:
5 g
500 g
2.5 kg
Packaging:
Glass bottle
Plastic bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC130760050 5 g Glass bottle
AC130765000 500 g Plastic bottle
AC130760025 2.5 kg Plastic bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC130760050 Supplier Thermo Scientific Chemicals Supplier No. 130760050
Only null left
Add to Cart

CAS: 90-43-7 | C12H10O | 170.21 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 90-43-7
Molecular Formula C12H10O
Molecular Weight (g/mol) 170.21
MDL Number MFCD00002208
InChI Key LLEMOWNGBBNAJR-UHFFFAOYSA-N
Synonym 2-hydroxybiphenyl, o-phenylphenol, biphenyl-2-ol, 2-biphenylol, o-hydroxybiphenyl, 2-hydroxydiphenyl, o-hydroxydiphenyl, phenylphenol, biphenylol, 1,1'-biphenyl-2-ol
PubChem CID 7017
ChEBI CHEBI:17043
SMILES OC1=CC=CC=C1C1=CC=CC=C1

Specifications

CAS Min % 99.0
CAS Max % 100.0
Melting Point >56.0°C
Color White
Density 1.2100g/mL
Boiling Point 282.0°C
Flash Point 123°C
Infrared Spectrum Authentic
Assay Percent Range 99% min. (GC)
Packaging Glass bottle
Linear Formula C6H5C6H4OH
Quantity 5 g
Beilstein 06,II,623
Specific Gravity 1.21
Merck Index 15,7415
Solubility Information Solubility in water: 0.7g/L(20°C). Other solubilities: readily soluble in naoh solution,ethanol and acet,800g/L isopropanol,soluble in most common organic solvents
IUPAC Name [1,1'-biphenyl]-2-ol
Viscosity 2.4 mPa.s (100°C)
Formula Weight 170.21
Percent Purity 99+%
Physical Form Crystalline Flakes
Chemical Name or Material 2-Phenylphenol, 99+%
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.
Very toxic to aquatic life.
Precautionary Statement GHS P Statement:
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Avoid release to the environment.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 201-993-5
RTECSNumber DV5775000
TSCA TSCA

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.