Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Phenylpiperidine 98.0+%, TCI America™
SDP

Catalog No. P20055G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P20055G 5 g
P20051G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P20055G Supplier TCI America Supplier No. P20055G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 3466-80-6
Molecular Formula C11H15N
Molecular Weight (g/mol) 161.248
MDL Number MFCD00270125
InChI Key WGIAUTGOUJDVEI-UHFFFAOYSA-N
Synonym 2-phenyl-piperidine, piperidine, 2-phenyl, 2-phenyl piperidine, pubchem8019, piperidin-2-yl benzene, acmc-1ag1g
PubChem CID 103020
IUPAC Name 2-phenylpiperidine
SMILES C1CCNC(C1)C2=CC=CC=C2

Specifications

Color Yellow
Boiling Point 112°C
Quantity 5 g
Formula Weight 161.25
Percent Purity ≥98.0% (GC,T)
Physical Form Liquid
Chemical Name or Material 2-Phenylpiperidine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.