Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich 2-Phenylpropionic acid

Catalog No. P3170110G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 g
10 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P3170110G 10 g
P317015G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P3170110G Supplier Sigma Aldrich Supplier No. P3170110G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

  • Tandem mass spectrometry data independently generated by scripps center for metabolomics is available to view or download in pdf
  • P31701.pdf tested metabolites are featured on scripps center for metabolomics metlin metabolite database
  • To learn more, visit sigma.com/metlin

Specifications

CAS 492-37-5
Density 1.1 g/mL (at 25°C (literature))
Boiling Point 260°C to 262°C (lit.)
Molecular Formula C9H10O2
Refractive Index n20/D 1.522 (literature)
Linear Formula CH3CH(C6H5)CO2H
MDL Number MFCD00002650
Quantity 10 g
Synonym (+/-)-2-Phenylpropionic acid; (+/-)-Hydratropic acid
Molecular Weight (g/mol) 150.17
Percent Purity 97%
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.