Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 2-phenylpropionic acid | 492-37-5 | MFCD00002650 | >98.0% | 150.17 g/mol | C9H10O2 | 10 G
SDP

Catalog No. 5000300669
Encompass_Preferred

2-Phenylpropionic acid (CAS 492-37-5) is an aromatic carboxylic acid used as a research reagent and chemical intermediate in synthetic chemistry and analytical applications. It is commonly supplied reagent-grade for laboratory use.

  • High purity: >98%.
  • Molecular weight: 150.17 g/mol.
  • Chemical formula: C9H10O2.
  • Physical properties: melting point ≈5 °C, boiling point 260-262 °C, density 1.1 g/mL.
  • Typical uses: synthetic intermediate and analytical reference standard.
  • Available in multiple pack sizes suitable for research use.

Catalog No. 50-003-00669 Supplier Medchemexpress LLC Supplier No. HYW01560810G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

2-Phenylpropionic acid (CAS 492-37-5) is an aromatic carboxylic acid used as a research reagent and chemical intermediate in synthetic chemistry and analytical applications. It is commonly supplied reagent-grade for laboratory use.

  • High purity: >98%.
  • Molecular weight: 150.17 g/mol.
  • Chemical formula: C9H10O2.
  • Physical properties: melting point ≈5 °C, boiling point 260-262 °C, density 1.1 g/mL.
  • Typical uses: synthetic intermediate and analytical reference standard.
  • Available in multiple pack sizes suitable for research use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.