Learn More
Medchemexpress LLC 2-phenylpropionic acid | 492-37-5 | MFCD00002650 | 99.7% | 150.18 g/mol | C9H10O2 | 1 G
SDP
Catalog No. 5000300068
Encompass_Preferred
2-Phenylpropionic acid (CAS 492-37-5) is an analytical standard and research chemical described as an intermediate in alpha-methylstyrene metabolism and a liver protein binder. It is used in studies of metabolism, protein-binding assays, and research related to nonsteroidal anti-inflammatory agents.
  • High purity suitable for analytical and research use.
  • Available in multiple package sizes for lab flexibility.
  • Provided as solid and DMSO solutions for convenient handling.
  • Well characterized with confirmed molecular weight and chemical formula.
  • Appropriate for metabolism and protein-binding studies.
Catalog No. 50-003-00068 Supplier Medchemexpress LLC Supplier No. HYW0156081G
2-Phenylpropionic acid (CAS 492-37-5) is an analytical standard and research chemical described as an intermediate in alpha-methylstyrene metabolism and a liver protein binder. It is used in studies of metabolism, protein-binding assays, and research related to nonsteroidal anti-inflammatory agents.
  • High purity suitable for analytical and research use.
  • Available in multiple package sizes for lab flexibility.
  • Provided as solid and DMSO solutions for convenient handling.
  • Well characterized with confirmed molecular weight and chemical formula.
  • Appropriate for metabolism and protein-binding studies.