Learn More
Medchemexpress LLC 2-Phenylpropionic acid | 492-37-5 | MFCD00002650 | 99.7% | 150.18 g·mol⁻¹ | C9H10O2 | 1 ML
SDP
Catalog No. 5000300731
Encompass_Preferred
2-Phenylpropionic acid is a small-molecule carboxylic acid (CAS 492-37-5) used as an analytical standard and synthetic intermediate in laboratory research. It is supplied as a ready-to-use 10 mM solution in DMSO and as bulk solid packs, providing a convenient source for assays, method development, and synthesis workflows.
  • High purity suitable for analytical and research use.
  • Offered as a ready-to-use 10 mM solution in DMSO for direct application.
  • Also available in solid quantities for synthesis and bulk use.
  • Molecular formula C9H10O2 and molecular weight 150.18 g·mol⁻¹ for precise calculations.
  • Compatible with common analytical techniques, including GC and LC-MS.
Catalog No. 50-003-00731 Supplier Medchemexpress LLC Supplier No. HYW01560810MM/1ML
2-Phenylpropionic acid is a small-molecule carboxylic acid (CAS 492-37-5) used as an analytical standard and synthetic intermediate in laboratory research. It is supplied as a ready-to-use 10 mM solution in DMSO and as bulk solid packs, providing a convenient source for assays, method development, and synthesis workflows.
  • High purity suitable for analytical and research use.
  • Offered as a ready-to-use 10 mM solution in DMSO for direct application.
  • Also available in solid quantities for synthesis and bulk use.
  • Molecular formula C9H10O2 and molecular weight 150.18 g·mol⁻¹ for precise calculations.
  • Compatible with common analytical techniques, including GC and LC-MS.