Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 2-phenylpropionic acid | 492-37-5 | MFCD00002650 | 99.7% | 150.18 g/mol | C9H10O2 | 25 G
SDP

Catalog No. 5000303455
Encompass_Preferred

2-Phenylpropionic acid (CAS 492-37-5) is a small-molecule analytical standard and metabolic intermediate used in research on alpha-methylstyrene metabolism and liver protein binding. It is intended for analytical and biochemical applications and is supplied with supporting documentation for quality control.

  • High reported purity (99.7%) for reliable analytical results.
  • Available in multiple pack sizes, including 25 g, for flexible usage scales.
  • Molecular formula C9H10O2 and molecular weight 150.18 g/mol for reference.
  • Documentation available: data sheet, SDS, and certificate of analysis.
  • Recommended storage conditions provided to maintain stability.

Catalog No. 50-003-03455 Supplier Medchemexpress LLC Supplier No. 25GHYW015608
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

2-Phenylpropionic acid (CAS 492-37-5) is a small-molecule analytical standard and metabolic intermediate used in research on alpha-methylstyrene metabolism and liver protein binding. It is intended for analytical and biochemical applications and is supplied with supporting documentation for quality control.

  • High reported purity (99.7%) for reliable analytical results.
  • Available in multiple pack sizes, including 25 g, for flexible usage scales.
  • Molecular formula C9H10O2 and molecular weight 150.18 g/mol for reference.
  • Documentation available: data sheet, SDS, and certificate of analysis.
  • Recommended storage conditions provided to maintain stability.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.