Learn More
Selleck Chemical LLC 2-Phenylpropionic acid
SDP
Catalog No. 5000003099
Encompass_Preferred
2-Phenylpropionic acid (2-Phenylpropionate Hydratropic acid -methyl- -toluic acid) is an intermediate in alpha-Methylstyrene (2-phenylpropylene) metabolism
Catalog No. 50-000-03099 Supplier Selleck Chemical LLC Supplier No. S632125UL
2-Phenylpropionic acid (2-Phenylpropionate Hydratropic acid -methyl- -toluic acid) is an intermediate in alpha-Methylstyrene (2-phenylpropylene) metabolism