Learn More
AdipoGen 2-Phenylpropionyl chloride
SDP
Catalog No. 502560223
Encompass_Preferred
Chemical. CAS 22414-26-2. Formula C9H9ClO. MW 168.6. Synthetic. Building block for synthesis.
Catalog No. 50-256-0223 Supplier AdipoGen Supplier No. CDXP0136G005
Chemical. CAS 22414-26-2. Formula C9H9ClO. MW 168.6. Synthetic. Building block for synthesis.