Learn More
Description
- Ethyl 3-methyl-2-oxobutyrate reacts with a variety of allyl halides in the presence of indium to afford hydroxy unsaturated carbonyl compounds
- Conformational behaviour of ethyl 3-methyl-2-oxobutyrate was investigated using solution ftir in combination with ab initio calculations.ethyl 3-methyl-2-oxobutyrate was used in the synthesis of (2sr,4sr)-5(s)-(n-boc)-amino-6-cyclohexyl-4-hydoxy-2-isopropyl hexanoic acid
Specifications
Specifications
| Molecular Formula | C10H8N2 |
| Linear Formula | C10H8N2 |
| MDL Number | MFCD04038761 |
| Quantity | 5 g |
| Molecular Weight (g/mol) | 156.18 |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.