Learn More
eMolecules​ 2-Phenylpyrimidine | Combi-Blocks | 7431-45-0 | MFCD04038761 | 156.188 | C10H8N2 | 95.000 | c1ccc(cc1)-c1ncccn1 | 5g | 232317430
Supplier:  eMolecules​ 232317430
Encompass_Preferred
2-Phenylpyrimidine | Combi-Blocks | 7431-45-0 | MFCD04038761 | 156.188 | C10H8N2 | 95.000 | c1ccc(cc1)-c1ncccn1 | 5g | 232317430
Catalog No. NC1676028