Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-(Phenylsulfonyl)acetophenone, 99%, Thermo Scientific™

Catalog No. AAL0503206
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0503206 5 g
AAL0503214 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL0503206 Supplier Thermo Scientific Chemicals Supplier No. L0503206

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3406-03-9

Specifications

Melting Point 92°C to 96°C
Molecular Formula C10H14N2O4
MDL Number MFCD00010570
Quantity 5 g
Beilstein 1879574
Synonym 3'-deoxythymidine, 2',3'-dideoxythymidine, 1-2r,5s-5-hydroxymethyl tetrahydrofuran-2-yl-5-methylpyrimidine-2,4 1h,3h-dione, dideoxythymidine, thymidine, 3'-deoxy, desoxythymidin, 1-2r,5s-5-hydroxymethyl tetrahydrofuran-2-yl-5-methyl-pyrimidine-2,4-dione, ddt & gm-csf
InChI Key XKKCQTLDIPIRQD-JGVFFNPUSA-N
SMILES CC1=CN([C@H]2CC[C@@H](CO)O2)C(=O)NC1=O
IUPAC Name 1-[(2R,5S)-5-(hydroxymethyl)oxolan-2-yl]-5-methylpyrimidine-2,4-dione
Molecular Weight (g/mol) 226.23
PubChem CID 65119
Formula Weight 260.31
Percent Purity 99%
Chemical Name or Material 2-(Phenylsulfonyl)acetophenone
Show More Show Less
Hazard Category H315-H319-H335
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 222-292-0
TSCA No
Recommended Storage Ambient temperatures
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.