Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
2-(Phenylthio)acetamidine hydrochloride, 98%, Thermo Scientific™
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
Specifications
| Melting Point | 175°C to 177°C |
| Quantity | 1 g |
| Formula Weight | 202.71 |
| Percent Purity | 98% |
| Chemical Name or Material | 2-(Phenylthio)acetamidine hydrochloride |
Safety and Handling
| Hazard Category | Warning |
| Hazard Statement | GHS H Statement H319 Causes serious eye irritation. |
| Precautionary Statement | GHS P Statement P280-P264-P305+P351+P338-P337+P313 Wear protective gloves/protective clothing/eye protection/face protection. Wash thoroughly after handling. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. IF eye irritation persists: Get medical advice/attention. |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?