Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-(Phenylthio)ethanethioamide, 97%, Thermo Scientific™

Catalog No. KM04192DA
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
KM04192DA 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. KM04192DA Supplier Thermo Scientific Chemicals Supplier No. KM04192DA

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 59865-82-6
Molecular Formula C8H9NS2
Molecular Weight (g/mol) 183.29
MDL Number MFCD00085154
InChI Key ZRDOSZLCJHRADP-UHFFFAOYSA-N
Synonym 2-phenylthio ethanethioamide, 2-phenylsulfanyl ethanethioamide, maybridge1_004803, 2-phenylthio thioacetamide, 2-phenylsulfanyl-thioacetamide, ethanethioamide,2-phenylthio, 1-amino-2-phenylthioethane-1-thione
PubChem CID 2778855
SMILES NC(=S)CSC1=CC=CC=C1

Specifications

CAS Min % 95.0
Assay Percent Range 95%
Packaging Amber glass bottle
Linear Formula C6H5SCH2CSNH2
Quantity 1 g
IUPAC Name 2-phenylsulfanylethanethioamide
Formula Weight 183.3
Percent Purity 95%
Chemical Name or Material 2-(Phenylthio)ethanethioamide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.