Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich 2-(Phenylthio)ethanol

Catalog No. 23277750G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
250 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
23-277-750G 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 23-277-750G Supplier Sigma Aldrich Supplier No. 23277750G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

2-(Phenylthio)ethanol

Specifications

CAS 699-12-7
Density 1.143 g/mL (at 25°C (literature))
Boiling Point 115°C to 116°C (2 mmHg)
Molecular Formula C8H10OS
Refractive Index n20/D 1.592 (literature)
Linear Formula C6H5SCH2CH2OH
MDL Number MFCD00002905
Quantity 250 mg
Synonym 2-Hydroxyethyl phenyl sulfide
Molecular Weight (g/mol) 154.23
Percent Purity 99%
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.