Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-(Phenylthio)ethanol 97.0+%, TCI America™
SDP

Catalog No. P073325ML Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P073325ML 25 mL
P0733500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P073325ML Supplier TCI America Supplier No. P073325ML
Add to Cart
Add to Cart

Chemical Identifiers

CAS 699-12-7
Molecular Formula C8H10OS
Molecular Weight (g/mol) 154.23
MDL Number MFCD00002905
InChI Key KWWZHCSQVRVQGF-UHFFFAOYSA-N
Synonym 2-phenylthio ethanol, phenylthioethanol, ethanol, 2-phenylthio, 2-hydroxyethyl phenyl sulfide, 2-phenylmercaptoethanol, 2-phenylsulfanyl ethanol, 2-phenylthioethanol, unii-9wa374o44p, pubchem10784
PubChem CID 69685
IUPAC Name 2-(phenylsulfanyl)ethan-1-ol
SMILES OCCSC1=CC=CC=C1

Specifications

Color Yellow
Boiling Point 275°C
Quantity 25 mL
Formula Weight 154.23
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material 2-(Phenylthio)ethanol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.