Learn More
Medchemexpress LLC HY-107837 500mg Medchemexpress, 2-Phospho-L-ascorbic acid (trisodium) CAS:66170-10-3 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY107837500MG
Encompass_Preferred
Medchemexpress, HY-107837 500mg 2-Phospho-L-ascorbic acid (trisodium) CAS:66170-10-3 2-Phospho-L-ascorbic acid trisodium acts as an antioxidant and a stimulator of hepatocyte growth factor (HGF) production. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-8475