Learn More
Medchemexpress LLC 2-phospho-L-ascorbic acid magnesium | 113170-55-1 | MFCD08063372 | 100.0% | 292.56 g/mol | C6H9O9P·3/2Mg | 100 MG
SDP
Catalog No. 5000244453
Encompass_Preferred
L-Ascorbic acid 2-phosphate magnesium is a stable, long-acting vitamin C derivative supplied as a white to off-white solid. It is used in cell culture and tissue engineering to stimulate collagen formation and expression, and offers greater stability than free ascorbic acid.
  • Stimulates collagen formation and expression.
  • Stable magnesium salt form with improved shelf life.
  • High purity suitable for research applications.
  • White to off-white solid for easy handling and weighing.
  • Store at 4°C in a sealed container to maintain stability.
  • Packaged in 100 mg quantity for small-scale experiments.
Catalog No. 50-002-44453 Supplier Medchemexpress LLC Supplier No. HY103701A100MG
L-Ascorbic acid 2-phosphate magnesium is a stable, long-acting vitamin C derivative supplied as a white to off-white solid. It is used in cell culture and tissue engineering to stimulate collagen formation and expression, and offers greater stability than free ascorbic acid.
  • Stimulates collagen formation and expression.
  • Stable magnesium salt form with improved shelf life.
  • High purity suitable for research applications.
  • White to off-white solid for easy handling and weighing.
  • Store at 4°C in a sealed container to maintain stability.
  • Packaged in 100 mg quantity for small-scale experiments.