Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 2-Phosphoglycolic acid lithium salt | 13147-57-4 | MFCD00133770 | 98.0% | 156.03 (free acid) | 1 MG
SDP

Supplier:  Medchemexpress LLC HY138877A1MG

Encompass_Preferred

Phosphoglycolic acid lithium is a competitive inhibitor of phosphoglycerate mutase (PGAM)-B, an enzyme involved in the glycolytic pathway. It increases basal and fMLF-induced levels of G6P in intact neutrophils, leading to an increase in upstream intermediates and amplifying the respiratory oxidative burst.

  • Competitive inhibitor of phosphoglycerate mutase (PGAM)-B
  • Increases basal and fMLF-induced levels of G6P in intact neutrophils
  • Amplifies the respiratory oxidative burst
  • Purity of 98.0%
  • Molecular weight of 156.03 (free acid)
  • Soluble in H2O at ≥ 50 mg/mL

Catalog No. 50-003-52104


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.