Learn More
Medchemexpress LLC HY-N7138 10mg Medchemexpress, 2-(Phosphonooxy)benzoic acid CAS:6064-83-1 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYN713810MG
Encompass_Preferred
Medchemexpress, HY-N7138 10mg 2-(Phosphonooxy)benzoic acid CAS:6064-83-1 2-(Phosphonooxy)benzoic acid is a non-acetylated salicylic acid derivative which has the potential for inflammatory disease as well as in analgesic therapy. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-203-1620