Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich 2-Picolylamine

Catalog No. A65204100G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A65204100G 100 g
A6520425G 25 g
A652045G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. A65204100G Supplier Sigma Aldrich Supplier No. A65204100G
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

  • 2-picolylamine is a bihaptic amine that can be used:
  • As a key precursor to synthesize various ionic liquids through the formation of α-amino alcohols as intermediates
  • As a chelating ligand for the synthesis of complexes such as zinc picolylamine complex and cu(ii) picolylamine complex
  • To functionalize poly(styrene-co-maleic anhydride) (psma) resin to facilitate the adsorption of uranium from aqueous solution

Specifications

CAS 3731-51-9
Density 1.049 g/mL at 25°C (lit.)
Boiling Point 82°C to 85°°C (12 mmHg)
Molecular Formula C6H8N2
Refractive Index n20/D 1.544 (lit.)
Linear Formula C6H8N2
MDL Number MFCD00006360
Quantity 100 g
Synonym 2-(Aminomethyl)pyridine
Molecular Weight (g/mol) 108.14
Percent Purity 99%
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.